Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 3 | 654 | ||
NM_018212.4 | NP_060682.2 | 3 | 654 | |||
XM_005273187.2 | XP_005273244.1 | 4 | 694 | |||
XM_011544229.2 | XP_011542531.2 | 3 | 1113 | |||
XM_017001746.1 | XP_016857235.1 | 3 | 1860 | |||
XM_017001747.1 | XP_016857236.1 | 3 | 1113 | |||
XM_017001748.1 | XP_016857237.1 | 3 | 1113 | |||
XM_017001749.1 | XP_016857238.1 | 3 | 1113 | |||
XM_017001750.1 | XP_016857239.1 | 3 | 1113 | |||
XM_017001751.1 | XP_016857240.1 | 3 | 1002 | |||
XM_017001752.1 | XP_016857241.1 | 3 | 1113 | |||
XM_017001753.1 | XP_016857242.1 | 4 | 664 | |||
XM_017001754.1 | XP_016857243.1 | 3 | 608 | |||
XM_017001755.1 | XP_016857244.1 | 3 | 351 | |||
XM_017001756.1 | XP_016857245.1 | 3 | 351 | |||
XM_017001757.1 | XP_016857246.1 | 3 | 254 | |||
GenBank mRNA | AF519769.1 | - | 3 | 470 | ||
AK290179.1 | - | 3 | 598 | |||
AY345143.1 | - | 3 | 558 | |||
BC065238.1 | - | 2 | 72 | |||
BC095481.1 | - | 3 | 653 | 30348815 | ||
EU255274.1 | - | 2 | 205 | |||
KM214203.1 | - | 2 | 201 | |||
KM214204.1 | - | 2 | 201 |