Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 7 | 1616 | ||
NM_018212.4 | NP_060682.2 | 7 | 1616 | |||
XM_005273187.2 | XP_005273244.1 | 8 | 1656 | |||
XM_011544229.2 | XP_011542531.2 | 8 | 2132 | |||
XM_017001746.1 | XP_016857235.1 | 9 | 2891 | |||
XM_017001747.1 | XP_016857236.1 | 9 | 2144 | |||
XM_017001748.1 | XP_016857237.1 | 7 | 2075 | |||
XM_017001749.1 | XP_016857238.1 | 8 | 2132 | |||
XM_017001750.1 | XP_016857239.1 | 8 | 2033 | |||
XM_017001751.1 | XP_016857240.1 | 7 | 1964 | |||
XM_017001752.1 | XP_016857241.1 | 6 | 1964 | |||
XM_017001753.1 | XP_016857242.1 | 10 | 1695 | |||
XM_017001754.1 | XP_016857243.1 | 9 | 1639 | |||
XM_017001755.1 | XP_016857244.1 | 9 | 1382 | |||
XM_017001756.1 | XP_016857245.1 | 7 | 1313 | |||
XM_017001757.1 | XP_016857246.1 | 7 | 1216 | |||
GenBank mRNA | AF519769.1 | - | 7 | 1432 | ||
AK001635.1 | - | 1 | 96 | |||
AK096246.1 | - | 5 | 912 | |||
AK126894.1 | - | 2 | 373 | |||
AK127561.1 | - | 1 | 747 | |||
AK290179.1 | - | 7 | 1560 | |||
AY345143.1 | - | 7 | 1520 | |||
BC065238.1 | - | 6 | 1034 | |||
BC095481.1 | - | 7 | 1615 | 30348815 | ||
EU255274.1 | - | 5 | 1056 | |||
KM214203.1 | - | 7 | 1220 | |||
KM214204.1 | - | 7 | 1220 |