Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 3 | 716 | ||
NM_018212.4 | NP_060682.2 | 3 | 716 | |||
XM_005273187.2 | XP_005273244.1 | 4 | 756 | |||
XM_011544229.2 | XP_011542531.2 | 3 | 1175 | |||
XM_017001746.1 | XP_016857235.1 | 3 | 1922 | |||
XM_017001747.1 | XP_016857236.1 | 3 | 1175 | |||
XM_017001748.1 | XP_016857237.1 | 3 | 1175 | |||
XM_017001749.1 | XP_016857238.1 | 3 | 1175 | |||
XM_017001750.1 | XP_016857239.1 | 3 | 1175 | |||
XM_017001751.1 | XP_016857240.1 | 3 | 1064 | |||
XM_017001752.1 | XP_016857241.1 | 3 | 1175 | |||
XM_017001753.1 | XP_016857242.1 | 4 | 726 | |||
XM_017001754.1 | XP_016857243.1 | 3 | 670 | |||
XM_017001755.1 | XP_016857244.1 | 3 | 413 | |||
XM_017001756.1 | XP_016857245.1 | 3 | 413 | |||
XM_017001757.1 | XP_016857246.1 | 3 | 316 | |||
GenBank mRNA | AF519769.1 | - | 3 | 532 | ||
AK096246.1 | - | 1 | 12 | |||
AK290179.1 | - | 3 | 660 | |||
AY345143.1 | - | 3 | 620 | |||
BC065238.1 | - | 2 | 134 | |||
BC095481.1 | - | 3 | 715 | 30348815 | ||
EU255274.1 | - | 2 | 267 | |||
KM214203.1 | - | 2 | 263 | |||
KM214204.1 | - | 2 | 263 |