Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161344.1 | NP_001154816.1 | 11 | 1379 | ||
NM_001161345.1 | NP_001154817.1 | 11 | 1379 | |||
NM_001161346.1 | NP_001154818.1 | 11 | 1343 | |||
NM_001161347.1 | NP_001154819.1 | 9 | 1103 | |||
NM_018223.2 | NP_060693.2 | 11 | 1256 | |||
GenBank mRNA | AF170724.1 | - | 11 | 1385 | ||
AK001658.1 | - | 11 | 1236 | |||
AK027687.1 | - | 11 | 1337 | |||
AK097671.1 | - | 9 | 1034 | |||
AK302785.1 | - | 9 | 1088 | |||
AK304333.1 | - | 11 | 1366 | |||
AL137561.1 | - | 221 | ||||
BC012072.1 | - | 10 | 1267 | 4650348 |