Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018238.3 | NP_060708.1 | 14 | 1180 | ||
XM_005250023.3 | XP_005250080.1 | 14 | 1076 | |||
XM_011516397.2 | XP_011514699.1 | 14 | 1646 | |||
GenBank mRNA | AJ278150.1 | - | 13 | 1036 | ||
AK001704.1 | - | 14 | 1048 | |||
AK222769.1 | - | 14 | 1042 | |||
AK299131.1 | - | 13 | 888 | |||
AK304502.1 | - | 15 | 1067 | |||
BC022777.1 | - | 13 | 1034 | 3913448 |