Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143996.1 | NP_001137468.1 | 10, 11 | 1445 | ||
NM_001143997.1 | NP_001137469.1 | 9, 10 | 1240 | |||
NM_018245.2 | NP_060715.2 | 11, 12 | 1616 | |||
XM_011539946.1 | XP_011538248.1 | 11, 12 | 1501 | |||
XM_011539947.1 | XP_011538249.1 | 11, 12 | 1707 | |||
XM_017016401.1 | XP_016871890.1 | 11, 12 | 1891 | |||
XM_017016402.1 | XP_016871891.1 | 11, 12 | 1518 | |||
XM_017016403.1 | XP_016871892.1 | 9, 10 | 1264 | |||
GenBank mRNA | AB033116.3 | - | 10, 11 | 1479 | ||
AK001713.1 | - | 11, 12 | 1560 | |||
AK290226.1 | - | 10, 11 | 1589 | |||
AK296551.1 | - | 10, 11 | 1354 | |||
AK303729.1 | - | 9, 10 | 1187 | |||
AK307759.1 | - | 10, 11 | 1375 | |||
BC026320.1 | - | 11, 12 | 1535 | 4797814 |