Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK5RAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011649.2 | NP_001011649.1 | 22 | 3030 | ||
NM_001272039.1 | NP_001258968.1 | 19 | 2340 | |||
NM_018249.5 | NP_060719.4 | 22 | 3030 | |||
NR_073554.1 | 22 | 3107 | ||||
NR_073555.1 | 22 | 3030 | ||||
NR_073556.1 | 23 | 3237 | ||||
NR_073557.1 | 22 | 3110 | ||||
NR_073558.1 | 22 | 3107 | ||||
XM_006717182.1 | XP_006717245.1 | 21 | 2923 | |||
XM_006717185.1 | XP_006717248.1 | 19 | 2332 | |||
XM_011518860.1 | XP_011517162.1 | 22 | 3016 | |||
XM_017014921.1 | XP_016870410.1 | 21 | 2920 | |||
XM_017014922.1 | XP_016870411.1 | 16 | 2248 | |||
XM_017014923.1 | XP_016870412.1 | 19 | 2332 | |||
XM_017014924.1 | XP_016870413.1 | 13 | 2004 | |||
GenBank mRNA | AB046853.2 | - | 22 | 2999 | ||
AF448860.1 | - | 22 | 2980 | |||
AK122913.1 | - | 22 | 3019 | |||
AL133161.1 | - | 6 | 1020 | |||
AL833481.1 | - | 22 | 3018 | |||
BC136275.1 | - | 22 | 2862 | 9020262 | ||
BC143762.1 | - | 22 | 2865 | 9052276 | ||
BC146782.1 | - | 22 | 2999 | 9007237 | ||
BX537421.1 | - | 22 | 3015 | |||
BX537708.1 | - | 19 | 2329 | |||
BX537759.1 | - | 22 | 3018 | |||
BX640896.1 | - | 15 | 2146 | |||
CR936622.1 | - | 15 | 2160 | |||
CR936717.1 | - | 15 | 2160 | |||
KP294527.1 | - | 22 | 3030 |