Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000084.4 | NP_000075.1 | 9 | 1700 | ||
NM_001127898.3 | NP_001121370.1 | 12 | 2044 | |||
NM_001127899.3 | NP_001121371.1 | 12 | 2303 | |||
NM_001282163.1 | NP_001269092.1 | 9 | 1743 | |||
XM_017029257.1 | XP_016884746.1 | 12 | 2006 | |||
XM_017029258.1 | XP_016884747.1 | 12 | 2315 | |||
GenBank mRNA | AK056560.1 | - | 12 | 1994 | ||
AK092049.1 | - | 12 | 2301 | |||
AK290940.1 | - | 9 | 1638 | |||
AK308089.1 | - | 9 | 1695 | |||
BC130429.1 | - | 8 | 1868 | 40146460 | ||
BC130431.1 | - | 8 | 1868 | 40146462 | ||
X91906.1 | - | 9 | 1743 |