Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCOR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136223.1 | NP_001129695.1 | 6 | 807 | ||
NM_001136224.2 | NP_001129696.2 | 6 | 807 | |||
NM_001136225.1 | NP_001129697.1 | 6 | 807 | |||
NM_018254.3 | NP_060724.1 | 5 | 607 | |||
XM_005273191.4 | XP_005273248.1 | 6 | 791 | |||
XM_006711442.2 | XP_006711505.1 | 6 | 791 | |||
XM_011509748.2 | XP_011508050.1 | 6 | 791 | |||
XM_011509749.2 | XP_011508051.1 | 6 | 791 | |||
XM_017001758.1 | XP_016857247.1 | 7 | 925 | |||
XM_017001759.1 | XP_016857248.1 | 7 | 900 | |||
XM_017001760.1 | XP_016857249.1 | 7 | 925 | |||
XM_017001761.1 | XP_016857250.1 | 6 | 807 | |||
XM_017001762.1 | XP_016857251.1 | 7 | 925 | |||
XM_017001763.1 | XP_016857252.1 | 7 | 925 | |||
XM_017001764.1 | XP_016857253.1 | 6 | 807 | |||
XM_017001765.1 | XP_016857254.1 | 7 | 925 | |||
XM_017001766.1 | XP_016857255.1 | 7 | 1135 | |||
XM_017001767.1 | XP_016857256.1 | 7 | 925 | |||
GenBank mRNA | AB037764.1 | - | 5 | 567 | ||
AK001738.1 | - | 5 | 579 | |||
AK131312.1 | - | 6 | 785 | |||
AK300942.1 | - | 7 | 873 | |||
AK302664.1 | - | 6 | 740 | |||
BC031608.1 | - | 6 | 659 | 5174915 |