Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018256.3 | NP_060726.3 | 4, 5 | 1092 | ||
GenBank mRNA | AF242546.1 | - | 4, 5 | 475 | ||
AK001743.1 | - | 4, 5 | 529 | |||
AK022781.1 | - | 4, 5 | 537 | |||
AK022782.1 | - | 4, 5 | 537 | |||
AK056092.1 | - | 4, 5 | 1092 | |||
BC008082.2 | - | 4, 5 | 472 | 3050219 | ||
EF011617.1 | - | 4, 5 | 471 |