Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024924.1 | NP_001020095.1 | 16 | 2369 | ||
NM_018261.3 | NP_060731.2 | 16 | 2213 | |||
NM_178237.2 | NP_839955.1 | 15 | 2168 | |||
XM_005265747.2 | XP_005265804.1 | 17 | 2457 | |||
XM_005265748.1 | XP_005265805.1 | 17 | 2269 | |||
XM_005265750.2 | XP_005265807.1 | 15 | 2245 | |||
XM_017008405.1 | XP_016863894.1 | 16 | 2412 | |||
XM_017008406.1 | XP_016863895.1 | 16 | 2224 | |||
XM_017008408.1 | XP_016863897.1 | 9 | 2467 | |||
XM_017008409.1 | XP_016863898.1 | 8 | 2422 | |||
GenBank mRNA | AF208854.1 | - | 7 | 840 | ||
AK001755.1 | - | 16 | 2155 | |||
AK023461.1 | - | 7 | 1927 | |||
AK027047.1 | - | 11 | 1316 | |||
AK027413.1 | - | 15 | 2085 | |||
AL833208.1 | - | 14 | 1956 | |||
BC019237.1 | - | 2 | 321 | |||
BC020650.1 | - | 7 | 837 | |||
BC094751.1 | - | 16 | 2369 | 4828034 |