Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFT122 SIRNA Assays ›
Gene Symbol:
IFT122
Gene Name:
intraflagellar transport 122
Gene Aliases:
CED, CED1, SPG, WDR10, WDR10p, WDR140
Chromosome Location:
Chr.3: 129440036 - 129520507 on Build GRCh38
UniGene ID:
Hs.655284
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31455
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IFT122
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001280541.1
NP_001267470.1
18
2444
NM_001280545.1
NP_001267474.1
19
2619
NM_001280546.1
NP_001267475.1
16
2212
NM_018262.3
NP_060732.2
17
2291
NM_052985.3
NP_443711.2
19
2621
NM_052989.2
NP_443715.1
18
2468
NM_052990.2
NP_443716.1
15
2135
XM_005247609.1
XP_005247666.1
17
2256
XM_006713689.1
XP_006713752.1
18
2433
XM_006713691.2
XP_006713754.1
17
2379
XM_006713692.2
XP_006713755.1
16
2202
XM_006713695.2
XP_006713758.1
18
2433
XM_011512972.2
XP_011511274.1
18
2433
XM_017006817.1
XP_016862306.1
18
2433
XM_017006818.1
XP_016862307.1
18
2433
XM_017006819.1
XP_016862308.1
17
2379
XM_017006820.1
XP_016862309.1
17
2379
XM_017006821.1
XP_016862310.1
17
2379
XM_017006822.1
XP_016862311.1
16
2224
XM_017006823.1
XP_016862312.1
17
2256
XM_017006824.1
XP_016862313.1
17
2256
XM_017006825.1
XP_016862314.1
16
2202
XM_017006826.1
XP_016862315.1
16
2202
XM_017006827.1
XP_016862316.1
16
2202
XM_017006828.1
XP_016862317.1
15
2047
XM_017006829.1
XP_016862318.1
15
2047
XM_017006830.1
XP_016862319.1
19
2431
XM_017006831.1
XP_016862320.1
17
2352
XM_017006832.1
XP_016862321.1
18
2331
XM_017006833.1
XP_016862322.1
14
1993
XM_017006834.1
XP_016862323.1
14
1993
XM_017006835.1
XP_016862324.1
16
2175
XM_017006836.1
XP_016862325.1
17
2200
XM_017006837.1
XP_016862326.1
9
1377
XM_017006838.1
XP_016862327.1
14
1817
GenBank mRNA
AF244930.1
-
18
2365
AF244931.1
-
19
2518
AF302154.1
-
18
2250
AK001759.1
-
17
2104
AK001955.1
-
7
913
AK022525.1
-
17
2202
AK023000.1
-
17
1933
AK024435.1
-
16
1979
AK094922.1
-
2
334
AK122965.1
-
11
1969
AK124140.1
-
19
2422
AK223095.1
-
17
2103
AK293852.1
-
14
1895
AK298526.1
-
14
1712
AK299903.1
-
16
1925
AK307463.1
-
5
698
AK315103.1
-
18
2268
AL133565.1
-
16
2045
AL833851.1
-
7
915
BC003045.1
-
15
1927
2823536
BC004238.2
-
15
1927
2823536
BC028353.2
-
18
2208
4836844
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
signal transduction downstream of smoothened
embryonic body morphogenesis
negative regulation of smoothened signaling pathway involved in ventral spinal cord patterning
embryonic heart tube development
embryonic forelimb morphogenesis
intraciliary anterograde transport
intraciliary retrograde transport
intraciliary transport involved in cilium morphogenesis
cilium assembly
embryonic digit morphogenesis
negative regulation of smoothened signaling pathway
camera-type eye morphogenesis
negative regulation of epithelial cell proliferation
limb development
cilium morphogenesis
ciliary receptor clustering involved in smoothened signaling pathway
smoothened signaling pathway involved in dorsal/ventral neural tube patterning
embryonic heart tube left/right pattern formation
protein localization to cilium
establishment of protein localization to organelle
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0