Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105549.1 | NP_001099019.1 | 6 | 2058 | ||
NM_001105550.1 | NP_001099020.1 | 5 | 1942 | |||
NM_001105551.1 | NP_001099021.1 | 5 | 1968 | |||
NM_001105552.1 | NP_001099022.1 | 4 | 1844 | |||
NM_001277945.1 | NP_001264874.1 | 6 | 2112 | |||
NM_001277946.1 | NP_001264875.1 | 4 | 1897 | |||
NM_001277947.1 | NP_001264876.1 | 6 | 2057 | |||
NM_001277948.1 | NP_001264877.1 | 5 | 1985 | |||
NM_001277949.1 | NP_001264878.1 | 5 | 2024 | |||
NM_001277951.1 | NP_001264880.1 | 3 | 1768 | |||
NM_001277952.1 | NP_001264881.1 | 5 | 1962 | |||
NM_018300.3 | NP_060770.3 | 3 | 1717 | |||
XM_017026951.1 | XP_016882440.1 | 7 | 2214 | |||
GenBank mRNA | AB209516.1 | - | 2 | 2857 | ||
AK001877.1 | - | 2 | 1333 | |||
AK027518.1 | - | 3 | 1762 | |||
AK027782.1 | - | 3 | 1498 | |||
AK055318.1 | - | 2 | 2249 | |||
AK075066.1 | - | 7 | 1935 | |||
AK225945.1 | - | 1 | 1468 | |||
AK289399.1 | - | 5 | 1718 | |||
AY500361.1 | - | 1 | 517 | |||
BC032867.1 | - | 7 | 1957 | 5264072 | ||
BC050407.1 | - | 1 | 1458 | 6197570 | ||
BC063032.1 | - | 2 | 1606 | 5553802 | ||
BG741860.1 | - | 1 | 595 | |||
BX647540.1 | - | 2 | 3574 | |||
M27877.1 | - | 1 | 1063 |