Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105549.1 | NP_001099019.1 | 6 | 813 | ||
NM_001105550.1 | NP_001099020.1 | 5 | 697 | |||
NM_001105551.1 | NP_001099021.1 | 5 | 723 | |||
NM_001105552.1 | NP_001099022.1 | 4 | 599 | |||
NM_001277945.1 | NP_001264874.1 | 6 | 867 | |||
NM_001277946.1 | NP_001264875.1 | 4 | 652 | |||
NM_001277947.1 | NP_001264876.1 | 6 | 812 | |||
NM_001277948.1 | NP_001264877.1 | 5 | 740 | |||
NM_001277949.1 | NP_001264878.1 | 5 | 779 | |||
NM_001277951.1 | NP_001264880.1 | 3 | 523 | |||
NM_001277952.1 | NP_001264881.1 | 5 | 717 | |||
NM_018300.3 | NP_060770.3 | 3 | 472 | |||
XM_017026951.1 | XP_016882440.1 | 7 | 969 | |||
GenBank mRNA | AB209516.1 | - | 2 | 1612 | ||
AK001877.1 | - | 1 | 172 | |||
AK027518.1 | - | 3 | 517 | |||
AK027782.1 | - | 2 | 337 | |||
AK055318.1 | - | 2 | 1004 | |||
AK075066.1 | - | 6 | 774 | |||
AK225945.1 | - | 1 | 223 | |||
AK289399.1 | - | 4 | 557 | |||
BC032867.1 | - | 6 | 796 | 5264072 | ||
BC050407.1 | - | 1 | 213 | 6197570 | ||
BC063032.1 | - | 2 | 361 | 5553802 | ||
BX647540.1 | - | 2 | 2329 | |||
DA258790.1 | - | 4 | 554 | |||
DA380756.1 | - | 1 | 181 | |||
DA731895.1 | - | 3 | 517 | |||
DR761375.1 | - | 2 | 410 |