Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018303.5 | NP_060773.3 | 5 | 676 | ||
NR_073064.1 | 7 | 838 | ||||
XM_017011018.1 | XP_016866507.1 | 6 | 722 | |||
XM_017011019.1 | XP_016866508.1 | 6 | 884 | |||
XM_017011020.1 | XP_016866509.1 | 6 | 744 | |||
XM_017011021.1 | XP_016866510.1 | 5 | 980 | |||
XM_017011022.1 | XP_016866511.1 | 5 | 676 | |||
XM_017011023.1 | XP_016866512.1 | 5 | 676 | |||
XM_017011024.1 | XP_016866513.1 | 6 | 722 | |||
XM_017011025.1 | XP_016866514.1 | 5 | 676 | |||
XM_017011026.1 | XP_016866515.1 | 5 | 676 | |||
GenBank mRNA | AJ414403.1 | - | 5 | 600 | ||
AK001888.1 | - | 1 | 91 | |||
AK056564.1 | - | 5 | 644 | |||
AK314628.1 | - | 5 | 676 | |||
AL833209.1 | - | 3 | 232 | |||
AL833213.1 | - | 5 | 622 | |||
BC016918.1 | - | 2 | 156 | |||
BC080542.1 | - | 7 | 808 | 5479496 |