Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018304.3 | NP_060774.2 | 7 | 1083 | ||
GenBank mRNA | AK000296.1 | - | 4 | 399 | ||
AK001891.1 | - | 7 | 836 | |||
AK225377.1 | - | 7 | 836 | |||
AK225380.1 | - | 7 | 841 | |||
BC008669.1 | - | 7 | 825 | 3867293 | ||
FJ161961.1 | - | 7 | 836 |