Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018368.3 | NP_060838.3 | 2 | 373 | ||
XM_011535941.1 | XP_011534243.1 | 2 | 223 | |||
GenBank mRNA | AF113224.1 | - | 2 | 93 | ||
AF208863.1 | - | 1 | 221 | |||
AK002102.1 | - | 2 | 122 | |||
AK225187.1 | - | 2 | 122 | |||
AK225537.1 | - | 2 | 107 | |||
AK290069.1 | - | 2 | 253 | |||
BC010360.1 | - | 2 | 105 | |||
BC047073.1 | - | 2 | 212 | 5274221 | ||
DA028310.1 | - | 2 | 373 |