Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEPDC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145208.1 | NP_001138680.1 | 2 | 198 | ||
NM_018369.2 | NP_060839.2 | 2 | 198 | |||
XM_011543509.2 | XP_011541811.1 | 2 | 191 | |||
XM_011543510.2 | XP_011541812.1 | 2 | 166 | |||
XM_017009629.1 | XP_016865118.1 | 2 | 166 | |||
GenBank mRNA | AF303178.1 | - | 2 | 166 | ||
AF490257.1 | - | 2 | 125 | |||
AK002114.1 | - | 2 | 166 | |||
AK290684.1 | - | 2 | 193 | |||
AK299242.1 | - | 3 | 232 | |||
AK300782.1 | - | 2 | 166 | |||
BC019075.1 | - | 2 | 161 | 5091386 | ||
DC299445.1 | - | 2 | 199 |