Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEPDC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145208.1 | NP_001138680.1 | 8, 9 | 1136 | ||
NM_018369.2 | NP_060839.2 | 8, 9 | 1136 | |||
XM_011543509.2 | XP_011541811.1 | 8, 9 | 1129 | |||
GenBank mRNA | AF303178.1 | - | 8, 9 | 1104 | ||
AF488828.1 | - | 5, 6 | 616 | |||
AF490257.1 | - | 8, 9 | 1063 | |||
AK001166.1 | - | 1, 2 | 47 | |||
AK002114.1 | - | 8, 9 | 1104 | |||
AK290684.1 | - | 8, 9 | 1131 | |||
AK299242.1 | - | 9, 10 | 1170 | |||
AK300782.1 | - | 8, 9 | 1104 | |||
BC010904.1 | - | 2, 3 | 204 | |||
BC019075.1 | - | 8, 9 | 1099 | 5091386 |