Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM63A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040217.2 | NP_001035307.1 | 5 | 902 | ||
NM_001163258.1 | NP_001156730.1 | 6 | 965 | |||
NM_001163259.1 | NP_001156731.1 | 5 | 867 | |||
NM_001163260.1 | NP_001156732.1 | 5 | 778 | |||
NM_001319998.1 | NP_001306927.1 | 6 | 2282 | |||
NM_018379.4 | NP_060849.2 | 6 | 1566 | |||
XM_005245321.3 | XP_005245378.1 | 5 | 1540 | |||
XM_005245325.3 | XP_005245382.1 | 5 | 895 | |||
XM_005245327.3 | XP_005245384.1 | 4 | 736 | |||
XM_005245328.3 | XP_005245385.1 | 4 | 736 | |||
XM_011509757.2 | XP_011508059.1 | 5 | 1540 | |||
XM_017001770.1 | XP_016857259.1 | 5 | 1540 | |||
XM_017001771.1 | XP_016857260.1 | 5 | 1278 | |||
XM_017001772.1 | XP_016857261.1 | 5 | 1278 | |||
XM_017001773.1 | XP_016857262.1 | 5 | 1540 | |||
XM_017001774.1 | XP_016857263.1 | 5 | 1278 | |||
XM_017001777.1 | XP_016857266.1 | 5 | 892 | |||
GenBank mRNA | AB037811.1 | - | 4 | 751 | ||
AK002142.1 | - | 5 | 628 | |||
AK125493.1 | - | 5 | 602 | |||
AK125959.1 | - | 6 | 2282 | |||
AK301946.1 | - | 6 | 965 | |||
AK303962.1 | - | 5 | 555 | |||
BC032321.1 | - | 5 | 1042 | 5240620 | ||
BC075812.1 | - | 6 | 1553 |