Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289107.1 | NP_001276036.1 | 8 | 1825 | ||
NM_018414.4 | NP_060884.1 | 7 | 1688 | |||
NR_110309.1 | 6 | 1584 | ||||
XM_011524995.2 | XP_011523297.1 | 7 | 1608 | |||
XM_011524996.2 | XP_011523298.1 | 7 | 1608 | |||
XM_017024842.1 | XP_016880331.1 | 7 | 1608 | |||
GenBank mRNA | AK000113.1 | - | 6 | 842 | ||
AY096001.1 | - | 7 | 1488 | |||
AY358918.1 | - | 7 | 1633 | |||
BC022462.1 | - | 6 | 1439 | 4816713 | ||
Y11339.2 | - | 7 | 1548 |