Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSNAXIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288990.1 | NP_001275919.1 | 3 | 489 | ||
NM_018430.3 | NP_060900.2 | 3 | 678 | |||
XM_005256049.4 | XP_005256106.1 | 3 | 794 | |||
XM_011523230.2 | XP_011521532.1 | 3 | 794 | |||
XM_011523231.2 | XP_011521533.1 | 3 | 794 | |||
XM_011523232.2 | XP_011521534.1 | 3 | 794 | |||
XM_011523233.2 | XP_011521535.1 | 3 | 794 | |||
XM_011523234.2 | XP_011521536.1 | 3 | 794 | |||
XM_011523235.1 | XP_011521537.1 | 3 | 429 | |||
XM_011523239.1 | XP_011521541.1 | 3 | 259 | |||
GenBank mRNA | AK027245.1 | - | 3 | 396 | ||
AK301920.1 | - | 3 | 262 | |||
BC111018.1 | - | 3 | 432 | 5168384 |