Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006579.2 | NP_006570.1 | 5 | 1112 | ||
GenBank mRNA | BC001549.1 | - | 5 | 1042 | 3463689 | |
BC001572.1 | - | 5 | 1044 | 3453423 | ||
BC046501.1 | - | 5 | 1086 | 5106272 | ||
Z37986.1 | - | 5 | 1018 |