Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289.5 | NP_001280.3 | 3 | 471 | ||
GenBank mRNA | AK292785.1 | - | 3 | 404 | ||
AY191592.1 | - | 4 | 271 | |||
BC005367.1 | - | 3 | 347 | |||
BC022305.1 | - | 3 | 347 | 4663624 | ||
BX482109.1 | - | 3 | 466 | |||
DA901964.1 | - | 3 | 471 | |||
Y12696.1 | - | 3 | 438 |