Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018448.4 | NP_060918.2 | 15 | 3950 | ||
GenBank mRNA | AB020636.2 | - | 15 | 3852 | ||
AF157326.1 | - | 15 | 3867 | |||
AK000976.1 | - | 4 | 503 | |||
AK001791.1 | - | 6 | 1351 | |||
AK027404.1 | - | 9 | 2585 | |||
AK292456.1 | - | 17 | 3789 | |||
AK314358.1 | - | 15 | 3838 | |||
AL133560.1 | - | 3 | 333 | |||
AL136810.1 | - | 15 | 3862 | |||
AL833880.2 | - | 9 | 2133 | |||
BC004232.1 | - | 6 | 1503 | |||
BC026220.2 | - | 13 | 3301 | |||
BC050341.1 | - | 15 | 3646 | 5265409 |