Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015691.3 | NP_056506.2 | 6 | 588 | ||
GenBank mRNA | AB033106.1 | - | 4 | 211 | ||
AK292961.1 | - | 5 | 470 | |||
AL353937.1 | - | 2 | 68 | |||
BC032447.1 | - | 3 | 123 | |||
BU500300.1 | - | 6 | 797 | |||
KC987947.1 | - | 6 | 765 |