Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288732.1 | NP_001275661.1 | 7 | 2841 | ||
NM_001288733.1 | NP_001275662.1 | 7 | 2833 | |||
NM_018469.4 | NP_060939.3 | 7 | 2862 | |||
XM_011524998.1 | XP_011523300.1 | 7 | 2851 | |||
XM_011524999.1 | XP_011523301.1 | 7 | 2775 | |||
XM_011525000.1 | XP_011523302.1 | 7 | 2832 | |||
XM_017024846.1 | XP_016880335.1 | 7 | 2811 | |||
GenBank mRNA | AB051525.2 | - | 6 | 2660 | ||
AF163260.1 | - | 5 | 878 | |||
AF220182.1 | - | 3 | 396 | |||
AK000369.1 | - | 2 | 167 | |||
AK091156.1 | - | 6 | 1830 | |||
AK091214.1 | - | 7 | 2758 | |||
AK314316.1 | - | 2 | 112 | |||
AL832371.1 | - | 7 | 2741 | |||
AL834251.2 | - | 7 | 2752 | |||
BC008896.2 | - | 6 | 2018 | |||
BC033661.1 | - | 4 | 544 | |||
BC036672.1 | - | 7 | 2812 | |||
BC040521.1 | - | 7 | 2766 | 4837394 | ||
BC040522.1 | - | 7 | 2814 | |||
BC108279.1 | - | 5 | 899 | |||
CR627433.1 | - | 7 | 2834 |