Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163022.1 | NP_001156494.1 | 10 | 1777 | ||
NM_001163023.1 | NP_001156495.1 | 9 | 1687 | |||
NM_001276389.1 | NP_001263318.1 | 9 | 1714 | |||
NM_018474.4 | NP_060944.3 | 11 | 1940 | |||
XM_011529296.2 | XP_011527598.1 | 11 | 1943 | |||
XM_011529297.2 | XP_011527599.1 | 11 | 1878 | |||
XM_011529298.2 | XP_011527600.1 | 11 | 1871 | |||
XM_011529299.2 | XP_011527601.1 | 10 | 1772 | |||
XM_017027951.1 | XP_016883440.1 | 12 | 1886 | |||
XM_017027952.1 | XP_016883441.1 | 10 | 1718 | |||
XM_017027953.1 | XP_016883442.1 | 12 | 1958 | |||
XM_017027954.1 | XP_016883443.1 | 10 | 1718 | |||
GenBank mRNA | AF220187.1 | - | 7 | 972 | ||
AF451990.1 | - | 11 | 1917 | |||
AK025476.1 | - | 11 | 1871 | |||
AK293166.1 | - | 9 | 1618 | |||
AK293567.1 | - | 9 | 1658 | |||
AK295355.1 | - | 10 | 1677 | |||
AK296399.1 | - | 7 | 1206 | |||
AL110196.1 | - | 7 | 1165 | |||
BC039296.1 | - | 11 | 1901 | |||
BC045826.1 | - | 10 | 1664 | 5262165 | ||
BC065550.1 | - | 9 | 1521 | |||
BC090879.2 | - | 7 | 1345 | |||
BC105093.1 | - | 11 | 1843 | 8144096 | ||
BC113370.1 | - | 11 | 1843 | 8322422 |