Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018476.3 | NP_060946.3 | 3 | 775 | ||
GenBank mRNA | AF183416.1 | - | 1 | 537 | ||
AF220189.1 | - | 3 | 701 | |||
AF237783.1 | - | 3 | 706 | |||
AY833561.1 | - | 3 | 697 | |||
BC018615.2 | - | 1 | 541 | |||
BM465948.1 | - | 3 | 716 |