Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166419.1 | NP_001159891.1 | 3 | 614 | ||
NM_001166420.1 | NP_001159892.1 | 3 | 614 | |||
NM_001166422.1 | NP_001159894.1 | 3 | 614 | |||
NM_018486.2 | NP_060956.1 | 3 | 614 | |||
NR_051952.1 | 3 | 614 | ||||
XM_011530986.2 | XP_011529288.2 | 3 | 609 | |||
XM_011530987.2 | XP_011529289.2 | 3 | 609 | |||
XM_017029640.1 | XP_016885129.1 | 3 | 609 | |||
XM_017029641.1 | XP_016885130.1 | 3 | 609 | |||
XM_017029643.1 | XP_016885132.1 | 3 | 609 | |||
XM_017029645.1 | XP_016885134.1 | 3 | 609 | |||
XM_017029647.1 | XP_016885136.1 | 3 | 609 | |||
GenBank mRNA | AF212246.1 | - | 3 | 330 | ||
AF230097.1 | - | 3 | 327 | |||
AF245664.1 | - | 3 | 281 | |||
AJ277724.1 | - | 3 | 314 | |||
AK074326.1 | - | 3 | 559 | |||
AK298767.1 | - | 3 | 354 | |||
AK300895.1 | - | 3 | 311 | |||
BC050433.1 | - | 3 | 305 | 5761745 | ||
BP229332.1 | - | 4 | 585 | |||
BQ189619.1 | - | 3 | 364 |