Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145355.1 | NP_001138827.1 | 9, 10 | 865 | ||
NM_001145356.1 | NP_001138828.1 | 8, 9 | 767 | |||
NM_018491.3 | NP_060961.3 | 8, 9 | 767 | |||
XM_005251513.4 | XP_005251570.1 | 7, 8 | 715 | |||
XM_005251517.4 | XP_005251574.1 | 8, 9 | 767 | |||
XM_011517961.2 | XP_011516263.1 | 9, 10 | 859 | |||
XM_011517962.2 | XP_011516264.1 | 8, 9 | 767 | |||
XM_011517963.1 | XP_011516265.1 | 5, 6 | 305 | |||
XM_017014925.1 | XP_016870414.1 | 8, 9 | 767 | |||
GenBank mRNA | AF212253.1 | - | 8, 9 | 735 | ||
AF257330.2 | - | 8, 9 | 725 | |||
AK290648.1 | - | 9, 10 | 837 | |||
AK297292.1 | - | 9, 10 | 806 | |||
AK315855.1 | - | 8, 9 | 691 | |||
AY343911.1 | - | 8, 9 | 724 | |||
BC005996.1 | - | 8, 9 | 767 | 4069589 | ||
BC009573.1 | - | 9, 10 | 818 | 4133536 | ||
BC013432.1 | - | 8, 9 | 738 | 4133722 | ||
BC132759.1 | - | 8, 9 | 701 | 40146781 |
Gene Symbol: CBWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291821.1 | NP_001278750.1 | 7, 8 | 688 | ||
NM_201453.3 | NP_958861.2 | 8, 9 | 748 | |||
XM_005277637.2 | XP_005277694.1 | 8, 9 | 827 | |||
XM_006717116.1 | XP_006717179.1 | 8, 9 | 827 | |||
XM_011518706.2 | XP_011517008.1 | 3, 4 | 278 | |||
XM_017014731.1 | XP_016870220.1 | 8, 9 | 3199 | |||
XM_017014733.1 | XP_016870222.1 | 3, 4 | 216 | |||
GenBank mRNA | AK297912.1 | - | 7, 8 | 650 | ||
AY344633.1 | - | 8, 9 | 724 |
Gene Symbol: CBWD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286835.1 | NP_001273764.1 | 9, 10 | 892 | ||
XM_005272748.1 | XP_005272805.1 | 8, 9 | 764 | |||
XM_005272749.1 | XP_005272806.1 | 8, 9 | 764 | |||
XM_005272750.1 | XP_005272807.1 | 7, 8 | 704 | |||
XM_006716914.1 | XP_006716977.1 | 8, 9 | 764 | |||
XM_017014456.1 | XP_016869945.1 | 9, 10 | 695 | |||
XM_017014458.1 | XP_016869947.1 | 8, 9 | 764 | |||
XM_017014459.1 | XP_016869948.1 | 10, 11 | 943 | |||
GenBank mRNA | AK097639.1 | - | 7, 8 | 1080 | ||
AK292585.1 | - | 9, 10 | 832 | |||
BC067803.1 | - | 9, 10 | 830 | 5294979 | ||
BC141856.1 | - | 9, 10 | 830 | |||
CD244615.1 | - | 1, 2 | 71 |