Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076552.2 | NP_001070020.2 | 2 | 432 | ||
NM_001242393.1 | NP_001229322.1 | 2 | 308 | |||
NM_018677.3 | NP_061147.1 | 2 | 432 | |||
XM_005260455.1 | XP_005260512.1 | 2 | 432 | |||
XM_005260456.1 | XP_005260513.1 | 2 | 432 | |||
XM_006723826.1 | XP_006723889.1 | 2 | 308 | |||
XM_011528905.1 | XP_011527207.1 | 2 | 385 | |||
XM_011528906.1 | XP_011527208.1 | 2 | 385 | |||
XM_011528907.1 | XP_011527209.1 | 2 | 385 | |||
XM_011528908.1 | XP_011527210.1 | 2 | 385 | |||
XM_011528909.2 | XP_011527211.1 | 2 | 271 | |||
XM_011528911.1 | XP_011527213.1 | 2 | 319 | |||
XM_011528912.2 | XP_011527214.1 | 2 | 277 | |||
GenBank mRNA | AF263614.1 | - | 2 | 311 | ||
AK000162.1 | - | 2 | 247 | |||
AK000188.1 | - | 2 | 385 | |||
AK025990.1 | - | 2 | 367 | |||
AK092281.1 | - | 2 | 344 | |||
AK098026.1 | - | 2 | 367 | |||
AK293634.1 | - | 2 | 364 | |||
AL359946.1 | - | 1 | 142 | |||
BC012172.1 | - | 2 | 355 | 4643236 | ||
BC073846.1 | - | 2 | 196 | |||
BC098422.1 | - | 1 | 27 | 5798672 | ||
CR749716.1 | - | 1 | 142 | |||
DA411343.1 | - | 2 | 432 | |||
DA626707.1 | - | 2 | 308 | |||
DA628275.1 | - | 3 | 320 | |||
DA630609.1 | - | 2 | 235 |