Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006493.2 | NP_006484.1 | 3, 4 | 733 | ||
GenBank mRNA | AF068227.1 | - | 3, 4 | 1997 | ||
AK075109.1 | - | 3, 4 | 573 | |||
AK304001.1 | - | 4, 5 | 488 |