Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006493.2 | NP_006484.1 | 2 | 371 | ||
XM_011534917.2 | XP_011533219.1 | 2 | 358 | |||
GenBank mRNA | AF068227.1 | - | 2 | 1635 | ||
AK075109.1 | - | 2 | 211 | |||
AK304001.1 | - | 3 | 126 |