Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161841.1 | NP_001155313.1 | 12, 13 | 2482 | ||
NM_018837.3 | NP_061325.1 | 12, 13 | 2143 | |||
NM_198596.2 | NP_940998.2 | 12, 13 | 2482 | |||
XM_005260458.2 | XP_005260515.1 | 12, 13 | 2058 | |||
XM_006723830.1 | XP_006723893.1 | 12, 13 | 2058 | |||
XM_011528916.1 | XP_011527218.1 | 12, 13 | 2495 | |||
XM_017027955.1 | XP_016883444.1 | 12, 13 | 2130 | |||
GenBank mRNA | AB033073.2 | - | 12, 13 | 2143 | ||
AK074483.1 | - | 6, 7 | 805 | |||
AK075035.1 | - | 5, 6 | 688 | |||
AL133001.1 | - | 1, 2 | 235 | |||
AY101176.1 | - | 12, 13 | 2482 | |||
AY358461.1 | - | 12, 13 | 2420 | |||
BC020962.1 | - | 1, 2 | 120 | |||
BC090952.1 | - | 1, 2 | 55 | |||
BC110539.1 | - | 11, 12 | 1909 | 40035480 | ||
CR749319.1 | - | 12, 13 | 2426 |