Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161841.1 | NP_001155313.1 | 7 | 1598 | ||
NM_018837.3 | NP_061325.1 | 7 | 1259 | |||
NM_198596.2 | NP_940998.2 | 7 | 1598 | |||
XM_005260458.2 | XP_005260515.1 | 7 | 1174 | |||
XM_006723827.1 | XP_006723890.1 | 7 | 1174 | |||
XM_006723828.1 | XP_006723891.1 | 7 | 1611 | |||
XM_006723829.2 | XP_006723892.1 | 7 | 1824 | |||
XM_006723830.1 | XP_006723893.1 | 7 | 1174 | |||
XM_011528913.1 | XP_011527215.1 | 7 | 1241 | |||
XM_011528914.1 | XP_011527216.1 | 7 | 1174 | |||
XM_011528915.1 | XP_011527217.1 | 7 | 1174 | |||
XM_011528916.1 | XP_011527218.1 | 7 | 1611 | |||
XM_017027955.1 | XP_016883444.1 | 7 | 1246 | |||
GenBank mRNA | AB033073.2 | - | 7 | 1259 | ||
AY101176.1 | - | 7 | 1598 | |||
AY358461.1 | - | 7 | 1536 | |||
BC110539.1 | - | 6 | 1025 | 40035480 | ||
CR749319.1 | - | 7 | 1542 |