Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019106.5 | NP_061979.3 | 2 | 321 | ||
NM_145733.2 | NP_663786.2 | 2 | 321 | |||
XM_017028862.1 | XP_016884351.1 | 3 | 1919 | |||
GenBank mRNA | AF285107.1 | - | 2 | 198 | ||
AF285108.1 | - | 2 | 198 | |||
AF285109.1 | - | 2 | 198 | |||
AK290037.1 | - | 2 | 306 | |||
AL833942.1 | - | 2 | 256 | |||
BC111779.1 | - | 2 | 214 | 40031828 | ||
BX465056.2 | - | 2 | 436 | |||
CR456572.1 | - | 2 | 210 |