Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC50A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122837.1 | NP_001116309.1 | 1 | 145 | ||
NM_001122839.1 | NP_001116311.1 | 1 | 145 | |||
NM_001287586.1 | NP_001274515.1 | 1 | 145 | |||
NM_001287587.1 | NP_001274516.1 | 1 | 145 | |||
NM_001287588.1 | NP_001274517.1 | 1 | 145 | |||
NM_001287589.1 | NP_001274518.1 | 1 | 145 | |||
NM_018845.3 | NP_061333.2 | 1 | 145 | |||
XM_006711453.2 | XP_006711516.1 | 1 | 145 | |||
GenBank mRNA | AF126023.1 | - | 1 | 105 | ||
AF126024.1 | - | 1 | 96 | |||
AK289551.1 | - | 1 | 146 | |||
BC005943.1 | - | 1 | 89 | 4076256 | ||
BC009621.1 | - | 1 | 107 | 3896154 | ||
BC010869.1 | - | 1 | 60 | |||
BC012378.1 | - | 1 | 94 | |||
BU160680.1 | - | 1 | 67 | |||
BU564973.1 | - | 1 | 132 | |||
CR457148.1 | - | 1 | 45 | |||
DA113832.1 | - | 1 | 146 | |||
X84712.1 | - | 1 | 16 |