Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031710.2 | NP_001026880.2 | 11 | 1804 | ||
NM_018846.4 | NP_061334.4 | 11 | 1888 | |||
NR_033328.1 | 12 | 1935 | ||||
XM_006715753.2 | XP_006715816.1 | 12 | 1838 | |||
XM_006715754.2 | XP_006715817.1 | 13 | 1921 | |||
XM_006715755.2 | XP_006715818.1 | 13 | 2140 | |||
XM_006715756.2 | XP_006715819.1 | 12 | 2009 | |||
XM_017012439.1 | XP_016867928.1 | 12 | 2101 | |||
GenBank mRNA | AK056390.1 | - | 12 | 1886 | ||
AK290479.1 | - | 11 | 1598 | |||
AK296219.1 | - | 8 | 1407 | |||
AK297369.1 | - | 1491 | ||||
AK297421.1 | - | 8 | 1328 | |||
AK297595.1 | - | 10 | 1645 | |||
AL136597.1 | - | 11 | 1750 | |||
AY028802.1 | - | 11 | 1453 | |||
BC039585.1 | - | 11 | 1746 | 5527346 | ||
EF560731.1 | - | 12 | 1654 |