Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016205.2 | NP_057289.1 | 5 | 1272 | ||
NR_036641.1 | 6 | 1333 | ||||
XM_011532124.2 | XP_011530426.1 | 7 | 952 | |||
XM_017008455.1 | XP_016863944.1 | 5 | 901 | |||
GenBank mRNA | AB033831.1 | - | 5 | 1107 | ||
AF091434.1 | - | 5 | 1272 | |||
AF244813.1 | - | 5 | 969 | |||
AF260738.1 | - | 5 | 943 | |||
AK300480.1 | - | 4 | 758 | |||
AK304638.1 | - | 7 | 615 | |||
AK313817.1 | - | 5 | 1002 | |||
AM922296.1 | - | 6 | 1153 | |||
AY358493.1 | - | 5 | 1065 | |||
BC094746.1 | - | 5 | 873 | |||
BC136662.1 | - | 5 | 1193 | 9020652 |