Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019109.4 | NP_061982.3 | 3 | 380 | ||
XM_011522565.1 | XP_011520867.1 | 3 | 187 | |||
XM_017023457.1 | XP_016878946.1 | 3 | 380 | |||
XM_017023458.1 | XP_016878947.1 | 3 | 502 | |||
GenBank mRNA | AB019038.1 | - | 3 | 354 | ||
AK075373.1 | - | 3 | 370 | |||
AK298144.1 | - | 3 | 187 | |||
AK311475.1 | - | 3 | 505 | |||
AY359073.1 | - | 3 | 364 | |||
BC004402.1 | - | 3 | 347 | 3510605 | ||
BC031095.1 | - | 3 | 359 | 5276692 | ||
BC031873.2 | - | 4 | 1106 | |||
BM767933.1 | - | 3 | 370 | |||
DA879477.1 | - | 3 | 381 |