Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018923.2 | NP_061746.1 | 1 | 1028 | ||
NM_032096.1 | NP_115267.1 | 1 | 1028 | |||
GenBank mRNA | AF152331.1 | - | 1 | 1028 | ||
AF152518.1 | - | 1 | 1028 | |||
BC101805.1 | - | 1 | 1091 | 8069311 |