Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018914.2 | NP_061737.1 | 1 | 1354 | ||
NM_032091.1 | NP_114480.1 | 1 | 1354 | |||
NM_032092.1 | NP_114481.1 | 1 | 1354 | |||
GenBank mRNA | AF152320.1 | - | 1 | 1096 | ||
AF152504.1 | - | 1 | 1096 | |||
AF152505.1 | - | 1 | 1096 | |||
BC136781.1 | - | 1 | 1129 | 9020771 | ||
BC171774.1 | - | 1 | 1129 | 9054428 |