Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018933.3 | NP_061756.1 | 1 | 1180 | ||
GenBank mRNA | AF152492.1 | - | 1 | 975 | ||
AF217745.1 | - | 1 | 975 | |||
AK292821.1 | - | 1 | 1142 | |||
AK296151.1 | - | 1 | 1143 | |||
AY358427.1 | - | 1 | 1145 | |||
BC033068.1 | - | 1 | 1133 | 5446016 | ||
BC051348.1 | - | 1 | 1133 | 6483735 | ||
BC071934.1 | - | 1 | 1133 | 6383780 |