Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018933.3 | NP_061756.1 | 1 | 461 | ||
GenBank mRNA | AF152492.1 | - | 1 | 256 | ||
AF217745.1 | - | 1 | 256 | |||
AK021915.1 | - | 1 | 444 | |||
AK292821.1 | - | 1 | 423 | |||
AK296151.1 | - | 1 | 424 | |||
AY358427.1 | - | 1 | 426 | |||
BC033068.1 | - | 1 | 414 | 5446016 | ||
BC051348.1 | - | 1 | 414 | 6483735 | ||
BC071934.1 | - | 1 | 414 | 6383780 | ||
DC329454.1 | - | 1 | 461 |
Gene Symbol: PCDHB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019120.4 | NP_061993.3 | 1 | 501 | ||
GenBank mRNA | AF152501.2 | - | 1 | 256 | ||
AU119840.1 | - | 1 | 444 | |||
BC136801.1 | - | 1 | 501 | 9020791 |