Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018898.3 | NP_061721.2 | 1 | 648 | ||
NM_031882.3 | NP_114088.2 | 1 | 648 | |||
GenBank mRNA | AF152303.1 | - | 1 | 472 | ||
AF152473.1 | - | 1 | 472 | |||
AJ007608.1 | - | 1 | 472 | |||
BC136774.1 | - | 1 | 493 | 9020764 | ||
BC136785.1 | - | 1 | 493 | 9020775 |