Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018902.4 | NP_061725.1 | 1 | 2587 | ||
NM_031861.2 | NP_114067.1 | 1 | 2587 | |||
GenBank mRNA | AF152307.1 | - | 1 | 996 | ||
AF152476.1 | - | 1 | 996 | |||
AJ007609.1 | - | 1 | 1853 | |||
BC136706.1 | - | 1 | 1917 | 9020696 |