Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018905.2 | NP_061728.1 | 1 | 975 | ||
NM_031495.1 | NP_113683.1 | 1 | 975 | |||
NM_031496.1 | NP_113684.1 | 1 | 975 | |||
GenBank mRNA | AF152310.1 | - | 1 | 869 | ||
AF152480.1 | - | 1 | 869 | |||
BC003126.1 | - | 1 | 975 | 3503666 |