Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260504.1 | NP_001247433.1 | 18 | 2558 | ||
NM_001260505.1 | NP_001247434.1 | 18 | 2282 | |||
NM_031414.4 | NP_113602.2 | 18 | 2282 | |||
NM_032944.3 | NP_116562.2 | 18 | 2526 | |||
NR_048542.1 | 18 | 2274 | ||||
XM_006715758.3 | XP_006715821.1 | 16 | 1662 | |||
XM_011515449.2 | XP_011513751.2 | 18 | 2208 | |||
XM_011515450.1 | XP_011513752.1 | 18 | 2529 | |||
XM_011515452.2 | XP_011513754.2 | 16 | 1675 | |||
GenBank mRNA | AF285599.1 | - | 18 | 2178 | ||
AF332194.1 | - | 18 | 2561 | |||
AK131214.1 | - | 9 | 984 | |||
AK292474.1 | - | 19 | 2174 | |||
AK302689.1 | - | 18 | 2174 | |||
BC033018.1 | - | 18 | 2176 | |||
BC059374.1 | - | 18 | 2189 | 5268051 |