Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260504.1 | NP_001247433.1 | 14 | 2213 | ||
NM_001260505.1 | NP_001247434.1 | 14 | 1937 | |||
NM_031414.4 | NP_113602.2 | 14 | 1937 | |||
NM_032944.3 | NP_116562.2 | 14 | 2181 | |||
NR_048542.1 | 14 | 1929 | ||||
XM_006715758.3 | XP_006715821.1 | 12 | 1317 | |||
XM_011515449.2 | XP_011513751.2 | 14 | 1863 | |||
XM_011515450.1 | XP_011513752.1 | 14 | 2184 | |||
XM_011515452.2 | XP_011513754.2 | 12 | 1330 | |||
GenBank mRNA | AF285599.1 | - | 14 | 1833 | ||
AF332194.1 | - | 14 | 2216 | |||
AK057271.1 | - | 14 | 1885 | |||
AK131214.1 | - | 5 | 639 | |||
AK292474.1 | - | 15 | 1829 | |||
AK302689.1 | - | 14 | 1829 | |||
BC033018.1 | - | 14 | 1831 | |||
BC059374.1 | - | 14 | 1844 | 5268051 |